Handheld refractometers
0 - 42 °Brix ; 42 - 71 °Brix ; 71 - 92 °Brix, 0,2 °Brix
Catalog # 635-0071
Supplier: EUROMEX
Jump to:
Product Details & Documents
Single dual or triple scales, analogue, handheld refractometers.
- Automated temperature compensation (ATC)
- Calibrated at 20 °C except for RF.5190
- Requires only a few drops of liquid
Specifications
- Range:0 - 42 °Brix ; 42 - 71 °Brix ; 71 - 92 °Brix
- Seq:08
- Precision:0,2 °Brix