Order Entry
Belgium
Orders LinkContactUsLinkComponent
Safe-Lock Tubes, 2,0 ml - Microtubes

Safe-Lock Tubes, 2,0 ml - Microtubes

2,0 ml, Colourless, Safe-Lock, PP, 25000×g, -, 1000 tubes
Catalog # 211-2120
   Low Manufacturing Impact
Supplier:   EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Catalog # 211-2120
Supplier:  EPPENDORF
   Low Manufacturing Impact
Safe-Lock Tubes, 2,0 ml - Microtubes
Safe-Lock Tubes, 2,0 ml - Microtubes

2,0 ml, Colourless, Safe-Lock, PP, 25000×g, -, 1000 tubes

Price
71,70 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Eppendorf® Safe-Lock 2,0 ml microtubes ensure secure sample preservation. With an innovative Safe-Lock mechanism, these microtubes prevent accidental openings, providing reliability in a 2,0 ml capacity. Ideal for various applications, they combine durability, ease of use, and sample protection.

  • Safe-Lock tube with a hinged lid prevents accidental opening during incubation and storage
  • g-Safe® technology, dependent on the Safe-Lock Tube variant, ensures exceptional centrifugation stability up to 30,000×g
  • g-Safe® technology prevents sample loss from tube breakage, and enhances safety when handling hazardous samples
  • Polypropylene construction imparts high resistance to chemicals, mechanical stress, and extreme temperatures
  • Precise lid sealing minimizes evaporation rates during prolonged storage
  • A large frosted lid and side surface for easy labelling
  • Functionality is assured within a temperature range from −86 to 100 °C
  • Autoclavable when open (121 °C, 20 min.) for effective sterilisation
  • Available in lot-certified PCR Clean, Eppendorf Forensic DNA Grade, and Biopur® quality for tested purity

Eppendorf® Safe-Lock 2,0 ml microtubes redefine secure sample storage. The patented Safe-Lock mechanism ensures protection against accidental openings, preserving sample integrity meticulously. Versatile across laboratory applications, the 2,0 ml capacity adapts to diverse sample volumes.

Constructed for durability, these microtubes withstand rigorous daily use. The user-friendly design, featuring clear graduations and a smooth surface, enhances visibility and handling ease. Elevate laboratory efficiency with Eppendorf® Safe-Lock tubes - trusted for reliability, security, and performance in sample management.

Manufactured without the use of slip agents, plasticisers, and biocides - substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.

Certifications: Certificate of Quality/Conformity, Lot-specific certficates for the purity grades PCR clean, Biopur®, Forensic DNA Grade can be downloaded from the Eppendorf website.

Specifications
  • Packed:
    1000 tubes
  • Sterile:
    -
  • Type:
    Safe-Lock
  • Capacity:
    2,0 ml
  • Colour:
    Colourless
  • Material:
    PP
  • Seq:
    02
  • RCF max.:
    25000×g
  • Cat. No.:
    211-2120
  • Operating temperature range:
    −86...100 °C
  • Environmental certification:
    Renewable Energy Certificate, ISO 14001
  • Volume:
    2,0 ml
  • Autoclavable:
    When open 121 °C, 20 min
  • Environmentally Preferable:
    Low Manufacturing Impact