Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer solutions

Buffer solutions

Buffer solution, 10,00, ±0,01, 20 °C, Plastic bottle, 5 L
  REGE5100
 :   REAGECON
Buffer solutions
  REGE5100
 :  REAGECON
 :  
Buffer solutions
Buffer solutions

Buffer solution, 10,00, ±0,01, 20 °C, Plastic bottle, 5 L

 :  
 
148,00 €
   
1 * 5 L
 

Documents

 

 

  • Pack type:
    Plastic bottle
  • Pk:
    5 L
  • Reference temperature:
    20 °C
  • Description:
    Buffer solution
  • pH accuracy:
    ±0,01
  • pH value:
    10,00

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.