Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer solutions

Buffer solutions

Buffer solution, 1,00, ±0,02, 20 °C, Plastic bottle, 5 L
Catalog # REGE5010
Supplier:   REAGECON
Buffer solutions
Catalog # REGE5010
Supplier:  REAGECON
CAS Number:  
Buffer solutions
Buffer solutions

Buffer solution, 1,00, ±0,02, 20 °C, Plastic bottle, 5 L

CAS Number:  
Price
164,00 €
Log in for availability
1 * 5 L
Product Details & Documents

 

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L
  • Reference temperature:
    20 °C
  • Description:
    Buffer solution
  • pH accuracy:
    ±0,02
  • pH value:
    1,00

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.