Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Buffer solutions

Buffer solution, 12,00, ±0,05, 20 °C, Plastic bottle, 1 L
  REGE1120
 :   REAGECON
Buffer solutions
  REGE1120
 :  REAGECON
 :  
Buffer solutions
Buffer solutions

Buffer solution, 12,00, ±0,05, 20 °C, Plastic bottle, 1 L

 :  
 
54,80 €
   
1 * 1 L
 

 

 

 

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Reference temperature:
    20 °C
  • Description:
    Buffer solution
  • pH accuracy:
    ±0,05
  • pH value:
    12,00