Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer solutions

Buffer solutions

Buffer solution, 10,00, ±0,01, 25 °C, Plastic bottle, 1 L
Catalog # REGE110025
Supplier:   REAGECON
Buffer solutions
Catalog # REGE110025
Supplier:  REAGECON
CAS Number:  
Buffer solutions
Buffer solutions

Buffer solution, 10,00, ±0,01, 25 °C, Plastic bottle, 1 L

CAS Number:  
Price
54,00 €
Log in for availability
1 * 1 L
Product Details & Documents

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Reference temperature:
    25 °C
  • Description:
    Buffer solution
  • pH accuracy:
    ±0,01
  • pH value:
    10,00

Specifications