Buffer solutions
Buffer solution, 9,00, ±0,01, 20 °C, Plastic bottle, 1 L
: REAGECON
- Type de conditionnement:Plastic bottle
- Cdt:1 L
- Température de réference:20 °C
- Désignation:Buffer solution
- Précision du pH:±0,01
- Valeur pH:9,00
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



