Buffer solutions
Buffer solution, 6,80, ±0,01, 25 °C, Plastic bottle, 500 mL
Catalog # REGE1068525
Supplier: REAGECON
Aller à :
Product Details & Documents
Documents
Certificats
Rechercher par numéro de lotSpécifications
Spécifications
- Pack type:Plastic bottle
- Pk:500 mL
- Reference temperature:25 °C
- Description:Buffer solution
- pH accuracy:±0,01
- pH value:6,80
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



