Orders LinkContactUsLinkComponent
Mechanical cell counters
Mechanical cell counters
Manual cell counter, 0 − 999, 8, Metal
Catalog # 720-0811
Supplier:   DENOMINATOR
Mechanical cell counters
Catalog # 720-0811
Supplier:  DENOMINATOR
Mechanical cell counters
Mechanical cell counters
Manual cell counter, 0 − 999, 8, Metal
Price
1 050,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Metal case mechanical cell counters that can count from 0 to 999.
  • Anti-slip strips
  • Reset button
  • Totalizer
Specifications
  • Range:
    0 − 999
  • Description:
    Manual cell counter
  • Material:
    Metal
  • Array:
    8