Orders LinkContactUsLinkComponent
Mechanical cell counters
Mechanical cell counters
Manual cell counter, 0 − 999, 2, Metal
Catalog # 720-0809
Supplier:   DENOMINATOR
Mechanical cell counters
Catalog # 720-0809
Supplier:  DENOMINATOR
Mechanical cell counters
Mechanical cell counters
Manual cell counter, 0 − 999, 2, Metal
Price
545,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Metal case mechanical cell counters that can count from 0 to 999.
  • Anti-slip strips
  • Reset button
  • Totalizer
Specifications
  • Range:
    0 − 999
  • Description:
    Manual cell counter
  • Material:
    Metal
  • Array:
    2
  • Cat. No.:
    720-0809