Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Calibration weights, OIML R111, class E1

1 g, 0,010 ± mg, Class E1 weights
  ZWIEE101.43.19
 :   ZWIEBEL
Calibration weights, OIML R111, class E1
  ZWIEE101.43.19
 :  ZWIEBEL
Calibration weights, OIML R111, class E1

1 g, 0,010 ± mg, Class E1 weights

 
 
   
1 * 1 ST
 
  • Weight:
    1 g
  • Seq:
    01
  • Tolerance:
    0,010 ± mg
  • Type:
    Class E1 weights
Accessories

 

  •  
    Description
     
     
  • ZWIE6570000
    Brush for cleaning weights, large, 200 mm length, flat, aluminium ferrule, 2 mm thick
       
    1 * 1 ST

 

  •  
    Description
     
     
  • ZWIE6930000
    Weight fork
       
    1 * 1 ST

 

  •  
    Description
     
     
  • ZWIEE660.00.00
    Gloves, cotton
       
    1 * 1 PAIRE

 

  •  
    Description
     
     
  • ZWIEE698.00.00
    Wooden box for single weight 1 g
       
    1 * 1 ST

 

  •  
    Description
     
     
  • ZWIEE698.00.06
    Wooden box for single weight 50 g
       
    1 * 1 ST

 

  •  
    Description
     
     
  • ZWIEE698.00.07
    Wooden box for single weight 100 g
       
    1 * 1 ST

 

  •  
    Description
     
     
  • ZWIEE698.00.08
    Wooden box for single weight 200 g
       
    1 * 1 ST

 

  •  
    Description
     
     
  • ZWIEE698.00.39
    Wooden box for single weight 500 g
       
    1 * 1 ST

 

  •  
    Description
     
     
  • ZWIEE699.00.02
    Plastic box for single weight 2 g
       
    1 * 1 ST

 

  •  
    Description
     
     
  • ZWIEE699.00.03
    Plastic box for single weight 5 g
       
    1 * 1 ST

 

  •  
    Description
     
     
  • ZWIEE699.00.07
    Plastic box for single weight 100 g
       
    1 * 1 ST