Orders LinkContactUsLinkComponent
Stages
Stages
Stage micrometer 25 +50/10 mm - 25 lines with 1 mm intervals in x-direction - 50 lines with 1/10 mm intervals in y-direction
Catalog # 630-0424
Supplier:   ZEISS
Stages
Catalog # 630-0424
Supplier:  ZEISS
Stages
Stage micrometer 25 +50/10 mm - 25 lines with 1 mm intervals in x-direction - 50 lines with 1/10 mm intervals in y-direction
Price
255,53 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Stage micrometer 25 +50/10 mm - 25 lines with 1 mm intervals in x-direction - 50 lines with 1/10 mm intervals in y-direction
  • Cat. No.:
    630-0424