Orders LinkContactUsLinkComponent
Objectives
Objectives
A-Planachromatic, N.A. of 0,45, WD of 0,51 mm
Catalog # 630-2250
Supplier:   ZEISS
Objectives
Catalog # 630-2250
Supplier:  ZEISS
Objectives
Objectives
A-Planachromatic, N.A. of 0,45, WD of 0,51 mm
Price
377,35 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Magnification:
    20×
  • Description:
    A-Planachromatic, N.A. of 0,45, WD of 0,51 mm
  • Seq:
    10