Order Entry
Belgium
Orders LinkContactUsLinkComponent
Halogen bulbs

Halogen bulbs

Halogen bulb with reflector, 8 V, 20 W
Catalog # 630-0333
Supplier:   ZEISS
Halogen bulbs
Catalog # 630-0333
Supplier:  ZEISS
Halogen bulbs

Halogen bulb with reflector, 8 V, 20 W

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Halogen bulb with reflector
  • Seq:
    13
  • Power:
    20 W
  • Voltage:
    8 V
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.