Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Halogen bulbs

Halogen bulb, 15 V, 150 W
  ZEIS4170530000000
 :   ZEISS
Halogen bulbs
  ZEIS4170530000000
 :  ZEISS
Halogen bulbs

Halogen bulb, 15 V, 150 W

 
17,89 €
   
1 * 1 ST
 
 
  • Description:
    Halogen bulb
  • Seq:
    12
  • Power:
    150 W
  • Voltage:
    15 V
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.