Orders LinkContactUsLinkComponent
Accessories for on-line multi-parameter systems, IQ sensor net
Accessories for on-line multi-parameter systems, IQ sensor net
VisoTurb® 700 IQ Digital turbidity sensor for potable water and waste water, with ultrasound cleaning system, without SACIQ cable
Catalog # 708-0049
Supplier:   WTW ONLINE
Accessories for on-line multi-parameter systems, IQ sensor net
Catalog # 708-0049
Supplier:  WTW ONLINE
Accessories for on-line multi-parameter systems, IQ sensor net
Accessories for on-line multi-parameter systems, IQ sensor net
VisoTurb® 700 IQ Digital turbidity sensor for potable water and waste water, with ultrasound cleaning system, without SACIQ cable
Price
3 700,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    VisoTurb® 700 IQ Digital turbidity sensor for potable water and waste water, with ultrasound cleaning system, without SACIQ cable
  • Seq:
    20