Orders LinkContactUsLinkComponent
Conductivity standards
Conductivity standards
E-Set Trace calibration set, 6 bottles with 50 ml calibration and control standard each, 0,01 mol/l KCl, 1413 µS/cm at 25 °C, accuracy ± 0.5 %, traceable to PTB/NIST, 1 SET
Catalog # 663-1891
Supplier:   WTW
Conductivity standards
Catalog # 663-1891
Supplier:  WTW
CAS Number:  
Conductivity standards
Conductivity standards
E-Set Trace calibration set, 6 bottles with 50 ml calibration and control standard each, 0,01 mol/l KCl, 1413 µS/cm at 25 °C, accuracy ± 0.5 %, traceable to PTB/NIST, 1 SET
CAS Number:  
Price
170,00 €
Log in for availability
1 * 1 SET
Specifications
Specifications
  • Pk:
    1 SET
  • Description:
    E-Set Trace calibration set, 6 bottles with 50 ml calibration and control standard each, 0,01 mol/l KCl, 1413 µS/cm at 25 °C, accuracy ± 0.5 %, traceable to PTB/NIST