Orders LinkContactUsLinkComponent
Accessories for on-line multi-parameter systems, IQ sensor net
Accessories for on-line multi-parameter systems, IQ sensor net
TriOxmatic 700 IQ Digital DO probe for waste water, with regeneration accessories, without SACIQ cable
Catalog # 708-0041
Supplier:   WTW ONLINE
Accessories for on-line multi-parameter systems, IQ sensor net
Catalog # 708-0041
Supplier:  WTW ONLINE
Accessories for on-line multi-parameter systems, IQ sensor net
Accessories for on-line multi-parameter systems, IQ sensor net
TriOxmatic 700 IQ Digital DO probe for waste water, with regeneration accessories, without SACIQ cable
Price
2 030,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    TriOxmatic 700 IQ Digital DO probe for waste water, with regeneration accessories, without SACIQ cable
  • Seq:
    12