Temperature probes
Temperature probe, with 1 m cable, water proof plug and built-in holding clip for pH electrodes, TFK 325/HC
Catalog # 662-1394
Supplier: WTW
Jump to:
Product Details & Documents
Specifications
- Description:Temperature probe, with 1 m cable, water proof plug and built-in holding clip for pH electrodes
- Type:TFK 325/HC
- Application:Automatic temperature compensation in aqueous solutions
Accessories
Product Information
- Catalog #AvailabilityPrice
- Holding clip for SenTix pH electrodes and temperature sensor TFK 325/HCHolding clip for SenTix pH electrodes and temperature sensor TFK 325/HCWTWA109821Log in for availability1 * 1 ST - 125,00 €