Orders LinkContactUsLinkComponent
Temperature probes
Temperature probes
Temperature probe, with 1 m cable and water proof plug, TFK 325
Catalog # 662-0264
Supplier:   WTW
Temperature probes
Catalog # 662-0264
Supplier:  WTW
Temperature probes
Temperature probes
Temperature probe, with 1 m cable and water proof plug, TFK 325
Price
205,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Temperature probe, with 1 m cable and water proof plug
  • Type:
    TFK 325
  • Application:
    Automatic temperature compensation in aqueous solutions
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Holding clip for SenTix pH electrodes and temperature sensor TFK 325/HC
    Holding clip for SenTix pH electrodes and temperature sensor TFK 325/HC
    WTWA109821
    Log in for availability
    1 * 1 ST - 125,00 €