Orders LinkContactUsLinkComponent
EasyPlus™ Titration Electrodes
EasyPlus™ Titration Electrodes
EM45-BNC sensor precipitation, ±2000 mV/0 - 14 pH, Easy Pro, Easy Cl titrators
Catalog # METT30043107
New Product
Supplier:   Mettler - Toledo
EasyPlus™ Titration Electrodes
Catalog # METT30043107
Supplier:  Mettler - Toledo
New Product
EasyPlus™ Titration Electrodes
EasyPlus™ Titration Electrodes
EM45-BNC sensor precipitation, ±2000 mV/0 - 14 pH, Easy Pro, Easy Cl titrators
Price
261,00 €
Log in for availability
1 * 1 Pce
Product Details & Documents
Electrode for use with Mettler Toledo® range of EasyPlus titrators only.
Electrode only.
Specifications
  • For:
    Easy Pro, Easy Cl titrators
  • Description:
    EM45-BNC sensor precipitation
  • Seq:
    3
  • Range:
    ±2000 mV/0 - 14 pH