EasyPlus™ Titration Electrodes
EG13-BNC sensor pH non-aqueous, 0 - 2000 mV, Easy Pro, Easy KFV, Easy Ox titrators
Catalog # METT30043104
New Product
Supplier: Mettler - Toledo
Jump to:
Product Details & Documents
Electrode for use with Mettler Toledo® range of EasyPlus™ titrators only.
Electrode only.
Specifications
- For:Easy Pro, Easy KFV, Easy Ox titrators
- Description:EG13-BNC sensor pH non-aqueous
- Seq:5
- Range:0 - 2000 mV