Orders LinkContactUsLinkComponent
EasyPlus™ Titration Electrodes
EasyPlus™ Titration Electrodes
EM40-BNC sensor redox mV, ±2000 mV, Easy Pro, Easy Ox titrators
Catalog # METT30043105
New Product
Supplier:   Mettler - Toledo
EasyPlus™ Titration Electrodes
Catalog # METT30043105
Supplier:  Mettler - Toledo
New Product
EasyPlus™ Titration Electrodes
EasyPlus™ Titration Electrodes
EM40-BNC sensor redox mV, ±2000 mV, Easy Pro, Easy Ox titrators
Price
315,00 €
Log in for availability
1 * 1 Pce
Product Details & Documents
Electrode for use with Mettler Toledo® range of EasyPlus titrators only.
Electrode only.
Specifications
  • For:
    Easy Pro, Easy Ox titrators
  • Description:
    EM40-BNC sensor redox mV
  • Seq:
    4
  • Range:
    ±2000 mV