Orders LinkContactUsLinkComponent
Blocks for COD
Blocks for COD
COD-W20-COMBO block, Ø 16,0/22,0 mm, 16/22 mm round-bottom tubes, 18/2, 47/45,5 mm
Catalog # 705-1208
New Product
Supplier:   VELP SCIENTIFIC
Blocks for COD
Catalog # 705-1208
Supplier:  VELP SCIENTIFIC
New Product
Blocks for COD
Blocks for COD
COD-W20-COMBO block, Ø 16,0/22,0 mm, 16/22 mm round-bottom tubes, 18/2, 47/45,5 mm
Price
136,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Dedicated to COD (Chemical Oxygen Demand) analysis in environmental testing, ensuring accurate digestion of water and wastewater samples under controlled conditions.
Specifications
  • No. of Holes:
    18/2
  • Description:
    COD-W20-COMBO block
  • Seq:
    2
  • Depth:
    47/45,5 mm
  • Well size:
    Ø 16,0/22,0 mm
  • For:
    16/22 mm round-bottom tubes