Orders LinkContactUsLinkComponent
Closures
Closures
Screw caps for HDPE white cans, OV 65, White, HDPE
Catalog # RIXI8-4110-65
New Product
Closures
Catalog # RIXI8-4110-65
New Product
Closures
Closures
Screw caps for HDPE white cans, OV 65, White, HDPE
Price
Inquire For Price
Log in for availability
1 * 200 ST
Product Details & Documents
Specifications
  • Thread:
    OV 65
  • Colour:
    White
  • Description:
    Screw caps for HDPE white cans
  • Material:
    HDPE
  • Seq:
    1