MATRIX MODIFIER SOLUTION PD(NO3)2 1 * 1 items
Catalog # REGEMMPD1050
New Product
Supplier: REAGECON
MATRIX MODIFIER SOLUTION PD(NO3)2 1 * 1 items
Catalog # REGEMMPD1050
Supplier: REAGECON
New Product
CAS Number:
Product Details & Documents
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



