Order Entry
Belgium
Orders LinkContactUsLinkComponent
Pipette tips

Pipette tips

Standard, 10000 µl, -, 164 mm, Red, Single tray (4 x 35)
Catalog # BHITLH-799000
New Product
Supplier:   Sartorius
Pipette tips
Catalog # BHITLH-799000
Supplier:  Sartorius
New Product
Pipette tips
Pipette tips

Standard, 10000 µl, -, 164 mm, Red, Single tray (4 x 35)

Price
129,00 €
Log in for availability
1 * 140 ST
Product Details & Documents

These pipette tips are suitable for many pipettes on the market.

  • Natural colour, premium grade PP, autoclavable
  • Excellent seal design for accurate, leak-tight sealing
  • Produced and packed in a dust-free environment under strictly maintained conditions

Ordering information: Available in bulk packaging, in autoclavable racks and in refills.

Caution: Article 613-4153 (BHIT780310) is designed for use with the Midi Plus Pipette Controller only.

Specifications
  • Colour code:
    Red
  • Packed:
    Single tray (4 x 35)
  • Sterile:
    -
  • Volume:
    10000 µl
  • Length:
    164 mm
  • Seq:
    45
  • Tip style:
    Standard
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.