Order Entry
Belgium
Orders LinkContactUsLinkComponent
2-Decyltetradecanol ≥98%

2-Decyltetradecanol ≥98%

Bottle
Catalog # APOSBUP13735-200MG
New Product
Supplier:   Apollo Scientific
2-Decyltetradecanol ≥98%
Catalog # APOSBUP13735-200MG
Supplier:  Apollo Scientific
New Product
CAS Number:  58670-89-6
2-Decyltetradecanol ≥98%

Bottle

CAS Number:  58670-89-6
Price
177,00 €
Log in for availability
1 * 200 mg
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  2-Decyltetradecan-1-ol
  • CAS Number:58670-89-6
  • Formula:C₂₄H₅₀O
  • Molecular Weight:354,65 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications