Dithranol ≥90%
Bottle
Catalog # APOSBUP14808-50MG
New Product
Supplier: Apollo Scientific
Dithranol ≥90%
Catalog # APOSBUP14808-50MG
Supplier: Apollo Scientific
New Product
CAS Number: 1143-38-0
Jump to:
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyms: 1,8,9-Trihydroxyanthracene, 1,8,9-Anthracenetriol, DIT, Dithranol, Anthranol, 1,8-Dihydroxyanthracen-9(10H)-one
- CAS Number:1143-38-0
- Formula:C₁₄H₁₀O₃
- Molecular Weight:226,23 g/mol
Specifications
Specifications
- Pack type:Bottle



