Order Entry
Belgium
Orders LinkContactUsLinkComponent
Dithranol ≥90%

Dithranol ≥90%

Bottle
Catalog # APOSBUP14808-200MG
New Product
Supplier:   Apollo Scientific
Dithranol ≥90%
Catalog # APOSBUP14808-200MG
Supplier:  Apollo Scientific
New Product
CAS Number:  1143-38-0
Dithranol ≥90%

Bottle

CAS Number:  1143-38-0
Price
81,10 €
Log in for availability
1 * 200 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  1,8,9-Trihydroxyanthracene, 1,8,9-Anthracenetriol, DIT, Dithranol, Anthranol, 1,8-Dihydroxyanthracen-9(10H)-one
  • CAS Number:1143-38-0
  • Formula:C₁₄H₁₀O₃
  • Molecular Weight:226,23 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications