Order Entry
Belgium
Orders LinkContactUsLinkComponent
3',4'-Dihydroxyacetophenone 98%

3',4'-Dihydroxyacetophenone 98%

Bottle
Catalog # APOSOR345013-100G
New Product
Supplier:   Apollo Scientific
3',4'-Dihydroxyacetophenone 98%
Catalog # APOSOR345013-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  1197-09-7
3',4'-Dihydroxyacetophenone 98%

Bottle

CAS Number:  1197-09-7
Price
257,00 €
Connexion Pour la disponibilité
1 * 100 g
Product Details & Documents
Synonymes:  1-(3,4-Dihydroxyphenyl)ethan-1-one, Acetopyrocatechol
  • Numéro CAS:1197-09-7
  • Formule:C₈H₈O₃
  • Numéro MDL:MFCD00016439
  • Poids moléculaire:152.15 g/mol
  • Température de stockage:Ambient
Spécifications

Spécifications

  • Pack type:
    Bottle

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.