Order Entry
Belgium
Orders LinkContactUsLinkComponent
3',4'-Dihydroxyacetophenone 98%

3',4'-Dihydroxyacetophenone 98%

Bottle
Catalog # APOSOR345013-25G
New Product
Supplier:   Apollo Scientific
3',4'-Dihydroxyacetophenone 98%
Catalog # APOSOR345013-25G
Supplier:  Apollo Scientific
New Product
CAS Number:  1197-09-7
3',4'-Dihydroxyacetophenone 98%

Bottle

CAS Number:  1197-09-7
Price
65,20 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  1-(3,4-Dihydroxyphenyl)ethan-1-one, Acetopyrocatechol
  • CAS Number:1197-09-7
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00016439
  • Molecular Weight:152.15 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.