Order Entry
Belgium
Orders LinkContactUsLinkComponent
Formaldehyde

Formaldehyde

Bottle
Catalog # APOSBIA0877-25ML
Flammable
Corrosive
Toxic
Health
New Product
Supplier:   Apollo Scientific
Formaldehyde
Catalog # APOSBIA0877-25ML
Supplier:  Apollo Scientific
New Product
CAS Number:  50-00-0
Formaldehyde

Bottle

CAS Number:  50-00-0
Price
86,90 €
Log in for availability
1 * 25 mL
Product Details & Documents
Synonyms:  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Boiling Pt:93…96 °C
  • Density:1.08…1.1 g/cm³ (25 °C)
  • Flash Pt:56 °C
  • Storage Temperature:Ambient
  • UN:2209
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Bottle

Specifications