Order Entry
Belgium
Orders LinkContactUsLinkComponent
α-Cyclodextrin

α-Cyclodextrin

Bottle
Catalog # APOSBIB6038-5G
Irritant
New Product
Supplier:   Apollo Scientific
α-Cyclodextrin
Catalog # APOSBIB6038-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  10016-20-3
α-Cyclodextrin

Bottle

CAS Number:  10016-20-3
Price
21,80 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:10016-20-3
  • Formula:C₃₆H₆₀O₃₀
  • MDL Number:MFCD00078207
  • Molecular Weight:972.84 g/mol
  • Melting Pt:>278 °C
  • Melting Pt:>278 °C
  • Storage Temperature:Ambient
  • EINECS:233-007-4
Specifications

Specifications

  • Pack type:
    Bottle

Specifications