Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tween® 20 (Polysorbate)

Tween® 20 (Polysorbate)

Bottle
Catalog # APOSBIT0803-250ML
New Product
Supplier:   Apollo Scientific
Tween® 20 (Polysorbate)
Catalog # APOSBIT0803-250ML
Supplier:  Apollo Scientific
New Product
CAS Number:  9005-64-5
Tween® 20 (Polysorbate)

Bottle

CAS Number:  9005-64-5
Price
36,20 €
Log in for availability
1 * 250 mL
Product Details & Documents

Non ionic detergent used for cell lysis, nuclei isolationand cell fractionation.

Synonyms:  Polyoxyethylene sorbitan monolaurate
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Molecular Weight:1227.53 g/mol
  • Boiling Pt:>100 °C
  • Melting Pt:–14 °C
  • Density:1.095 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:500-018-3

Documents

Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.