Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Cilostamide ≥95%

Bottle
  APOSBUP05815-100MG
New Product
Cilostamide ≥95%
  APOSBUP05815-100MG
New Product
 :  68550-75-4
Cilostamide ≥95%

Bottle

 :  68550-75-4
 
530,00 €
   
1 * 100 mg
 

Usage: Biochemical/life science applications

 :  N-Cyclohexyl-N-methyl-4-((2-oxo-1,2-dihydroquinolin-6-yl)oxy)butanamide
  •  :68550-75-4
  •  :C₂₀H₂₆N₂O₃
  •  :342,43 g/mol
 

 

  • Pack type:
    Bottle