Order Entry
Belgium
Orders LinkContactUsLinkComponent
3',4'-Dihydroxyacetophenone ≥98%

3',4'-Dihydroxyacetophenone ≥98%

Bottle
Catalog # APOSBUP18399-500MG
New Product
Supplier:   Apollo Scientific
3',4'-Dihydroxyacetophenone ≥98%
Catalog # APOSBUP18399-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  1197-09-7
3',4'-Dihydroxyacetophenone ≥98%

Bottle

CAS Number:  1197-09-7
Price
37,70 €
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  1-(3,4-Dihydroxyphenyl)ethan-1-one, Acetopyrocatechol
  • CAS Number:1197-09-7
  • Formula:C₈H₈O₃
  • Molecular Weight:152,15 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications