Py-BODIPY-NHS ester ≥98%
Bottle
Catalog # APOSBUP02580-50MG
New Product
Supplier: Apollo Scientific
Py-BODIPY-NHS ester ≥98%
Catalog # APOSBUP02580-50MG
Supplier: Apollo Scientific
New Product
CAS Number: 201998-61-0
Jump to:
Product Details & Documents
Usage: Biochemical/life science applications
Synonyms: Py-BODIPY-NHS ester, 2,5-Dioxopyrrolidin-1-yl 3-(5,5-difluoro-7-(1H-pyrrol-2-yl)-5H-5l4,6l4-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-3-yl)propanoate
- CAS Number:201998-61-0
- Formula:C₂₀H₁₇BF₂N₄O₄
- Molecular Weight:426,18 g/mol
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



