Order Entry
Belgium
Orders LinkContactUsLinkComponent
Creatinine ≥95%

Creatinine ≥95%

Bottle
Catalog # APOSBUP18477-500MG
New Product
Supplier:   Apollo Scientific
Creatinine ≥95%
Catalog # APOSBUP18477-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  60-27-5
Creatinine ≥95%

Bottle

CAS Number:  60-27-5
Price
53,60 €
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one
  • CAS Number:60-27-5
  • Formula:C₄H₇N₃O
  • Molecular Weight:113,12 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications