Order Entry
Belgium
Orders LinkContactUsLinkComponent
Erythromycin ≥98%

Erythromycin ≥98%

Bottle
Catalog # APOSBUP18313-5G
New Product
Supplier:   Apollo Scientific
Erythromycin ≥98%
Catalog # APOSBUP18313-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  114-07-8
Erythromycin ≥98%

Bottle

CAS Number:  114-07-8
Price
125,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  (-)-Erythromycin
  • CAS Number:114-07-8
  • Formula:C₃₇H₆₇NO₁₃
  • Molecular Weight:733,93 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.