Hydroxyectoin 95%
Bottle
Catalog # APOSOR1025890-250M
New Product
Supplier: Apollo Scientific
Hydroxyectoin 95%
Catalog # APOSOR1025890-250M
Supplier: Apollo Scientific
New Product
CAS Number: 165542-15-4
Jump to:
Product Details & Documents
Usage: Organic building block/reagent.
- CAS Number:165542-15-4
- Formula:C₆H₁₀N₂O₃
- Molecular Weight:158,16 g/mol
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



