Hydroxyectoin 95%
Bottle
Catalog # APOSOR1025890-100M
New Product
Supplier: Apollo Scientific
Hydroxyectoin 95%
Catalog # APOSOR1025890-100M
Supplier: Apollo Scientific
New Product
CAS Number: 165542-15-4
Jump to:
Product Details & Documents
Usage: Organic building block/reagent.
- CAS Number:165542-15-4
- Formula:C₆H₁₀N₂O₃
- Molecular Weight:158,16 g/mol
Specifications
Specifications
- Pack type:Bottle



