Vanillin 97+%
Bottle
Catalog # APOSOR52392-500G
New Product
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 4-Hydroxy-3-methoxybenzaldehyde
- CAS Number:121-33-5
- Formula:C₈H₈O₃
- MDL Number:MFCD00006942
- Molecular Weight:152.15 g/mol
- Boiling Pt:170 °C/15mmHg
- Melting Pt:81…83 °C
- Density:1.056 g/cm³ (25 °C)
- Flash Pt:153 °C
- Storage Temperature:Ambient
- EINECS:204-465-2
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Bottle



