Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Vanillin 97+%

Bottle
Catalog # APOSOR52392-100G
Irritant
New Product
Supplier:   Apollo Scientific
Vanillin 97+%
Catalog # APOSOR52392-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  121-33-5
Vanillin 97+%

Bottle

CAS Number:  121-33-5
Price
24,60 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  4-Hydroxy-3-methoxybenzaldehyde
  • CAS Number:121-33-5
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00006942
  • Molecular Weight:152.15 g/mol
  • Boiling Pt:170 °C/15mmHg
  • Melting Pt:81…83 °C
  • Density:1.056 g/cm³ (25 °C)
  • Flash Pt:153 °C
  • Storage Temperature:Ambient
  • EINECS:204-465-2

 

Specifications

Specifications

  • Pack type:
    Bottle

Specifications