Order Entry
Belgium
Orders LinkContactUsLinkComponent
Vanillin 97+%

Vanillin 97+%

Bottle
Catalog # APOSOR52392-25G
Irritant
New Product
Supplier:   Apollo Scientific
Vanillin 97+%
Catalog # APOSOR52392-25G
Supplier:  Apollo Scientific
New Product
CAS Number:  121-33-5
Vanillin 97+%

Bottle

CAS Number:  121-33-5
Price
21,80 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  4-Hydroxy-3-methoxybenzaldehyde
  • CAS Number:121-33-5
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00006942
  • Molecular Weight:152.15 g/mol
  • Boiling Pt:170 °C/15mmHg
  • Melting Pt:81…83 °C
  • Density:1.056 g/cm³ (25 °C)
  • Flash Pt:153 °C
  • Storage Temperature:Ambient
  • EINECS:204-465-2

Documents

Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.