MPC Coupling Caps
Product Details & Documents
MPC connectors are multi-purpose, open-format connectors for use in a wide variety of critical fluid handling applications. This particular type of connector is perfect for smaller bag systems for aliquoted media or other products stored in bags.
Ergonomic thumb latch: Easy to operate - even with gloved hands
- USP Class VI materials: Meet biocompatibility
- Sterilisable by autoclave, EtO, e-beam, or gamma: Robust and economical for single-use applications
- Parting line-free hose barb: Eliminates potential leak path
- BSE/TSE free materials: Meet ADCF requirements
MPC connectors add ease of use and security to the most critical fluid handling applications. Choose from a full line of male and female MPC connectors and configurations, including pressure sealing caps and plugs, in sizes to fit ¹/₈", ¹/₄", and ³/₈" tubing. MPC couplings offer optional locking sleeves to further guard against accidental disconnects. In addition, coupling halves can be rotated when connected to reduce tube kinks.
Specifications
- Description:SaniQuik USP class VI, ADCF silicone seal, 1-¹/₂"; 50 Pk
- Seq:10
- Fitting material:Silicone
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.