Order Entry
Belgium
Orders LinkContactUsLinkComponent
Copper 99,9%, nanopowder 30 - 50 nm

Copper 99,9%, nanopowder 30 - 50 nm

Supplier:   Apollo Scientific
Copper 99,9%, nanopowder 30 - 50 nm
Supplier:  Apollo Scientific
Certificates
Copper 99,9%, nanopowder 30 - 50 nm

4 Options Available Below

Product Details & Documents

Usage: Inorganic salt or catalyst

  • CAS Number:7440-50-8
  • Formula:Cu
  • MDL Number:MFCD00010965
  • Molecular Weight:63,546 g/mol
  • UN:3089
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 46,40 €
    46,40 €
    1 * 5 g
       46,40 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 109,00 €
    109,00 €
    1 * 25 g
       109,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 315,00 €
    315,00 €
    1 * 100 g
       315,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 1 790,00 €
    1 790,00 €
    1 * 500 g
       1 790,00 €

    Specifications:
    New Product
    New Product