Order Entry
Belgium
Orders LinkContactUsLinkComponent
Uridine ≥98%

Uridine ≥98%

New Product
Supplier:   Apollo Scientific
Uridine ≥98%
Supplier:  Apollo Scientific
New Product
Certificates

3 Options Available Below

Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside
  • CAS Number:58-96-8
  • Formula:C₉H₁₂N₂O₆
  • Molecular Weight:244,2 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 69,50 €
    69,50 €
    1 * 100 mg
       69,50 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 112,00 €
    112,00 €
    1 * 500 mg
       112,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 529,00 €
    529,00 €
    1 * 10 g
       529,00 €

    Specifications:
    New Product
    New Product