Order Entry
Belgium
Orders LinkContactUsLinkComponent
Ethidium bromide ≥98%

Ethidium bromide ≥98%

Bottle
Catalog # APOSBUP18019-250MG
New Product
Supplier:   Apollo Scientific
Ethidium bromide ≥98%
Catalog # APOSBUP18019-250MG
Supplier:  Apollo Scientific
New Product
CAS Number:  1239-45-8
Ethidium bromide ≥98%

Bottle

CAS Number:  1239-45-8
Price
53,60 €
Log in for availability
1 * 250 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
  • CAS Number:1239-45-8
  • Formula:C₂₁H₂₀BrN₃
  • Molecular Weight:394,32 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications