Order Entry
Belgium
Orders LinkContactUsLinkComponent
Docusate sodium ≥98%

Docusate sodium ≥98%

New Product
Supplier:   Apollo Scientific
Docusate sodium ≥98%
Supplier:  Apollo Scientific
New Product
Certificates
Docusate sodium ≥98%

4 Options Available Below

Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  Dioctyl sulphosuccinate sodium salt, Dioctyl sulphosuccinate, sodium salt, Sodium dioctyl sulphosuccinate, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulphonate, bis(2-Ethylhexyl) sodium sulphosuccinate
  • CAS Number:577-11-7
  • Formula:C₂₀H₃₇NaO₇S
  • Molecular Weight:444,56 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 63,70 €
    63,70 €
    1 * 100 mg
       63,70 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 95,60 €
    95,60 €
    1 * 200 mg
       95,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 158,00 €
    158,00 €
    1 * 500 mg
       158,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 523,00 €
    523,00 €
    1 * 5 g
       523,00 €

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.